Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ECM1, Mouse, Clone: SC05, Abnova™

Catalog No. 89109492 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-492 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-492 Supplier Abnova Corporation Supplier No. MAB11329
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against ECM1.

This gene encodes an extracellular protein containing motifs with a cysteine pattern characteristic of the cysteine pattern of the ligand-binding double-loop domains of the albumin protein family. This gene maps outside of the epidermal differentiation complex (EDC), a cluster of three gene families involved in epidermal differentiation. Alternatively spliced transcript variants encoding distinct isoforms have been described. [provided by RefSeq]

Specifications

Antigen ECM1
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone SC05
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against ECM1.
Dilution Western Blot (1:100-1:2000) Electron Microscopy (1:10-1:500) ELISA (1:100-1:2000) Immunocytochemistry/Immunofluorescence (1:10-1:2000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:500) Immunoprecipitation (1:10-1:
Formulation In PBS (0.05% BSA, 0.05% sodium azide)
Gene ECM1
Gene Symbols ECM1
Host Species Mouse
Immunogen 1989)., Partially purified ECM1 from human colostrum (Bartek
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1893
Test Specificity The antibody reacts with a reduction-resistant epitope present in both free and SIgA bound Secretory Component. However, the exact epitope recognized by the antibody has not been mapped.
Target Species Human, Rat
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.