Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ECs2841, Mouse, Clone: 744f8, Abnova™

Catalog No. 89113648 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-648 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-648 Supplier Abnova Corporation Supplier No. MAB8921
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant ECs2841.

Specifications

Antigen ECs2841
Applications ELISA, Western Blot
Classification Monoclonal
Clone 744f8
Concentration 1 ug/uL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant ECs2841.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In citrate-Tris-HCl, pH7.0 (0.02% Proclin 300)
Gene ECs2841
Gene Symbols ECs2841
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 23-338 of ECs2841.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 912508
Test Specificity This antibody recognizes recombinant protein RfbE.
Target Species E. coli
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.