Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDAR, Mouse, Clone: 3A7, Abnova™

Catalog No. 89106425 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-425 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-425 Supplier Abnova Corporation Supplier No. H00010913M02A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant EDAR.

This gene encodes a member of the tumor necrosis factor receptor family. The encoded transmembrane protein is a receptor for the soluble ligand ectodysplasin A, and can activate the nuclear factor-kappaB, JNK, and caspase-independent cell death pathways. It is required for the development of hair, teeth, and other ectodermal derivatives. Mutations in this gene result in autosomal dominant and recessive forms of hypohidrotic ectodermal dysplasia. [provided by RefSeq

Sequence: TKDEDYGCVPCPAEKFSKGGYQICRRHKDCEGFFRATVLTPGDMENDAECGPCLPGYYMLENRPRNIYGMVCYSCLLAPPNTKECVGATSGASANFPGTSGSSTLSPFQHAHKEL

Specifications

Antigen EDAR
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3A7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant EDAR.
Formulation ascites with no preservative
Gene EDAR
Gene Accession No. NM_022336
Gene Alias DL/ED1R/ED3/ED5/EDA-A1R/EDA1R/EDA3/FLJ94390
Gene Symbols EDAR
Host Species Mouse
Immunogen EDAR (NP_071731,64 a.a. ∽ 178 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10913
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.