Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDN1, Mouse, Clone: IC4, Abnova™

Catalog No. 89113134 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-134 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-134 Supplier Abnova Corporation Supplier No. MAB7146

Mouse monoclonal antibody raised against EDN1.

Specifications

Antigen EDN1
Applications Immunohistochemistry (Frozen), Immunoradiometric Assay
Classification Monoclonal
Clone IC4
Conjugate Unconjugated
Description endothelin 1
Formulation In PBS, pH 7.2 (0.09% sodium azide)
Gene EDN1
Gene Alias ET1, HDLCQ7
Gene Symbols EDN1
Host Species Mouse
Immunogen C-terminus of human EDN1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1906
Target Species Human
Content And Storage Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.