Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDN3, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104415 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-415 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-415 Supplier Abnova Corporation Supplier No. H00001908A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length recombinant EDN3.

The protein encoded by this gene is a member of the endothelin family. Endothelins are endothelium-derived vasoactive peptides involved in a variety of biological functions. The active form of this protein is a 21 amino acid peptide processed from the precursor protein. The active peptide is a ligand for endothelin receptor type B (EDNRB). The interaction of this endothelin with EDNRB is essential for development of neural crest-derived cell lineages, such as melanocytes and enteric neurons. Mutations in this gene and EDNRB have been associated with Hirschsprung disease (HSCR) and Waardenburg syndrome (WS), which are congenital disorders involving neural crest-derived cells. Four alternatively spliced transcript variants encoding three distinct isoforms have been observed. [provided by RefSeq

Sequence: MEPGLWLLFGLTVTSAAGFVPCSQSGDAGRRGVSQAPTAARSEGDCEETVAGPGEETVAGPGEGTVAPTALQGPSPGSPGQEQAAEGAPEHHRSRRCTCFTYKDKECVYYCHLDIIWINTPEQTVPYGLSNYRGSFRGKRSAGPLPGNLQLSHRPHLRCACVGRYDKACLHFCTQTLDVSSNSRTAEKTDKEEEGKVEVKDQQSKQALDLHHPKLMPGSGLALAPSTCPRCLFQEGAP

Specifications

Antigen EDN3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length recombinant EDN3.
Formulation 50% glycerol
Gene EDN3
Gene Accession No. BC008876
Gene Alias ET3/MGC15067/MGC61498
Gene Symbols EDN3
Host Species Mouse
Immunogen EDN3 (AAH08876, 1 a.a. to 238 a.a) full-length recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1908
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.