Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDNRA, Mouse, Clone: RJT24, Abnova™

Catalog No. 89136703 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
3 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-703 3 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-703 Supplier Abnova Corporation Supplier No. MAB9593

Mouse monoclonal antibody raised against partial recombinant EDNRA.

Specifications

Antigen EDNRA
Applications Immunohistochemistry
Classification Monoclonal
Clone RJT24
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant EDNRA.
Dilution Immunohistochemistry (1:50) The optimal working dilution should be determined by the end user.
Formulation In tissue culture supernatant (0.09% sodium azide)
Host Species Mouse
Immunogen Recombinant protein corresponding to a 55 amino acid fragment at C-terminus of EDNRA.
Quantity 3 x 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.