Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EEF2, Mouse, Clone: 5B6, Abnova™

Catalog No. 89108824 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-824 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-824 Supplier Abnova Corporation Supplier No. MAB10629
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant EEF2.

This gene encodes a member of the GTP-binding translation elongation factor family. This protein is an essential factor for protein synthesis. It promotes the GTP-dependent translocation of the nascent protein chain from the A-site to the P-site of the ribosome. This protein is completely inactivated by EF-2 kinase phosporylation. [provided by RefSeq

Specifications

Antigen EEF2
Applications ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 5B6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant EEF2.
Dilution ELISA (1:10000) Western Blot (1:500-1:2000) Immunofluorescence (1:200-1:1000) The optimal working dilution should be determined by the end user.
Formulation In ascites (0.03% sodium azide)
Gene EEF2
Gene Alias EEF-2/EF2
Gene Symbols EEF2
Host Species Mouse
Immunogen Recombinant protein corresponding to human EEF2.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1938
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.