Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EFEMP2, Mouse, Clone: 1C2, Abnova™

Catalog No. 89106815 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-815 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-815 Supplier Abnova Corporation Supplier No. H00030008M37
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant EFEMP2.

A large number of extracellular matrix proteins have been found to contain variations of the epidermal growth factor (EGF) domain and have been implicated in functions as diverse as blood coagulation, activation of complement and determination of cell fate during development. The protein encoded by this gene contains four EGF2 domains and six calcium-binding EGF2 domains. This gene is necessary for elastic fiber formation and connective tissue development. Defects in this gene are cause of an autosomal recessive cutis laxa syndrome. [provided by RefSeq

Sequence: SPQDSEEPDSYTECTDGYEWDPDSQHCRDVNECLTIPEACKGEMKCINHYGGYLCLPRSAAVINDLHGEGPPPPVPPAQHPNPCPPGYEPDDQDSCVDVDECAQALHDCRPSQDCHNLPGSYQCTCPDGYRKIGPECVDIDECRYRYCQHRCVNLPGSFRCQCEPGFQLGPNNRSCVDVNECDMGAPCEQRCFNSYGTFLCRCHQGYELHRDGFSCSDIDECSYSSYLCQYRCVNEPGRFSCHCPQGYQLLATRLCQDIDECESGAHQCSEAQTCVNFHGGYRCVDTNRCVEPYIQVSENRCLCPASNPLCREQPSSIVHRYMTITSERSVPADVFQIQATSVYPGAYNAFQIRAGNSQGDFYIRQINNVSAMLVLARPVTGPREYVLDLEMVTMNSLMSYRASSVLRLTVFVGAYTF

Specifications

Antigen EFEMP2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1C2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant EFEMP2.
Formulation PBS with no preservative; pH 7.4
Gene EFEMP2
Gene Accession No. BC010456.1
Gene Alias FBLN4/MBP1/UPH1
Gene Symbols EFEMP2
Host Species Mouse
Immunogen EFEMP2 (AAH10456.1,26 a.a. ∽ 443 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A Purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30008
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.