Learn More
EFEMP2, Mouse, Clone: 2C8, Abnova™
Mouse monoclonal antibody raised against a partial recombinant EFEMP2.
Supplier: Abnova Corporation H00030008M19
Description
A large number of extracellular matrix proteins have been found to contain variations of the epidermal growth factor (EGF) domain and have been implicated in functions as diverse as blood coagulation, activation of complement and determination of cell fate during development. The protein encoded by this gene contains four EGF2 domains and six calcium-binding EGF2 domains. This gene is necessary for elastic fiber formation and connective tissue development. Defects in this gene are cause of an autosomal recessive cutis laxa syndrome. [provided by RefSeq
Sequence: SPQDSEEPDSYTECTDGYEWDPDSQHCRDVNECLTIPEACKGEMKCINHYGGYLCLPRSAAVINDLHGEGPPPPVPPAQHPNPCPPGYEPDDQDSCVDVDECAQALHDCRPSQDCHNLPGSYQCTCPDGYRKIGPECVDIDECRYRYCQHRCVNLPGSFRCQCEPGFQLGPNNRSCVDVNECDMGAPCEQRCFNSYGTFLCRCHQGYELHRDGFSCSDIDECSYSSYLCQYRCVNEPGRFSCHCPQGYQLLATRLCQDIDECESGAHQCSEAQTCVNFHGGYRCVDTNRCVEPYIQVSENRCLCPASNPLCREQPSSIVHRYMTITSERSVPADVFQIQATSVYPGAYNAFQIRAGNSQGDFYIRQINNVSAMLVLARPVTGPREYVLDLEMVTMNSLMSYRASSVLRLTVFVGAYTFSpecifications
EFEMP2 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC010456 | |
EFEMP2 | |
EFEMP2 (AAH10456.1,26 a.a. ∽ 443 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Immunoprecipitation, Western Blot | |
2C8 | |
Mouse monoclonal antibody raised against a partial recombinant EFEMP2. | |
EFEMP2 | |
FBLN4/MBP1/UPH1 | |
Mouse | |
Protein A Purification | |
RUO | |
30008 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.