Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EFHD1, Mouse, Clone: 1F5, Abnova™
Mouse monoclonal antibody raised against a partial recombinant EFHD1.
Supplier: Abnova Corporation H00080303M08A
Description
EFHD1 is an EF-hand domain-containing protein that displays increased expression during neuronal differentiation (Tominaga and Tomooka, 2002 [PubMed 12270117]).[supplied by OMIM
Sequence: GLMALAKLSEIDVALEGVKGAKNFFEAKVQALSSASKFEAELKAEQDERKREEEERRLRQAAFQKLKANFNSpecifications
EFHD1 | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
NM_025202 | |
EFHD1 | |
EFHD1 (NP_079478.1,168 a.a. ∽ 238 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
80303 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |
ELISA, Western Blot | |
1F5 | |
Mouse monoclonal antibody raised against a partial recombinant EFHD1. | |
EFHD1 | |
DKFZp781H0842/FLJ13612/MST133/MSTP133/PP3051 | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction