Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGF, Mouse, Clone: KT57, Abnova™

Catalog No. 89113746 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-746 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-746 Supplier Abnova Corporation Supplier No. MAB9726
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant EGF.

Epidermal growth factor has a profound effect on the differentiation of specific cells in vivo and is a potent mitogenic factor for a variety of cultured cells of both ectodermal and mesodermal origin. The EGF precursor is believed to exist as a membrane-bound molecule which is proteolytically cleaved to generate the 53-amino acid peptide hormone that stimulates cells to divide. [provided by RefSeq

Specifications

Antigen EGF
Applications ELISA, Western Blot
Classification Monoclonal
Clone KT57
Concentration 1 mg/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant EGF.
Dilution ELISA (1 ug/mL) Sandwich ELISA (5 ug/mL for coating) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene EGF
Gene Alias HOMG4/URG
Gene Symbols EGF
Host Species Mouse
Immunogen Recombinant protein corresponding to human EGF.
Purification Method Protein A purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1950
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C to -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.