Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-EGF R/ErbB1, Clone: B1D8, Azide Free, Novus Biologicals™

Mouse Monoclonal Antibody
Specifications
Antigen | EGFR |
---|---|
Clone | B1D8 |
Concentration | 1.0 mg/mL |
Dilution | Flow Cytometry 0.5-1ug/million cells, Immunoprecipitation 1-2ug/500ug protein lysate, Blocking/Neutralizing, CyTOF-ready |
Applications | Flow Cytometry, Immunoprecipitation, Blocking Assay, CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP23455302
![]() |
Novus Biologicals
NBP2345530.2 MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
EGFR Monoclonal antibody specifically detects EGFR in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Flow Cytometry, Immunoprecipitation, CyTOF-ready, Block/Neutralize.Specifications
EGFR | |
1.0 mg/mL | |
Flow Cytometry, Immunoprecipitation, Blocking Assay, CyTOF | |
Unconjugated | |
Mouse | |
Cancer, Cell Biology, Cell Cycle and Replication, Growth and Development, Hypoxia, Phospho Specific, Signal Transduction, Tumor Suppressors, Tyrosine Kinases | |
10mM PBS - BSA free with No Preservative | |
avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ERBB, ErbB1, ERBB1PIG61, HER1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1 | |
EGFR | |
IgG2a κ | |
Protein A purified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
B1D8 | |
Flow Cytometry 0.5-1ug/million cells, Immunoprecipitation 1-2ug/500ug protein lysate, Blocking/Neutralizing, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse (Negative), Rat (Negative) | |
P00533 | |
1956 | |
Microsomes from A431 cells | |
Primary | |
Protein A purified | |
This antibody reacts with the extracellular domain of EGFR and blocks the EGF/TGF??induced activation. It also blocks tumor growth in vivo. It is excellent for purification of EGFR. EGFR is type I receptor tyrosine kinase with sequence homology to erbB-1, -2, -3 -4 or HER-1, -2, -3 -4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-a (TGF-a), Heparin-binding EGF (HB-EGF), amphiregulin, Beta cellulin and epiregulin. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title