Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Egg white avidin, Mouse, Clone: A4D12, Abnova™
Mouse monoclonal antibody raised against Egg white avidin.
Supplier: Abnova Corporation MAB2776
Specifications
Egg white avidin | |
Monoclonal | |
Unconjugated | |
Mouse | |
200 μg | |
Primary | |
Store at 4°C. | |
IgG1 |
ELISA, Immunohistochemistry, Western Blot | |
A4D12 | |
In PBS, pH 7.4 (0.09% sodium azide) | |
Egg white avidin. | |
RUO | |
This antibody specific to egg white avidin. No cross-reaction with human and animal serum proteins and streptavidin. React with avidin without interfering with its biotin-binding activity. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction