Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Egg white avidin, Mouse, Clone: A7B5, Abnova™

Catalog No. 89135227 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-227 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-227 Supplier Abnova Corporation Supplier No. MAB2777

Mouse monoclonal antibody raised against Egg white avidin.

Specifications

Antigen Egg white avidin
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone A7B5
Conjugate Unconjugated
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Egg white avidin.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody specific to egg white avidin. No cross-reaction with human and animal serum proteins and streptavidin. React with avidin without interfering with its biotin-binding activity.
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.