Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-EGR1, Clone: 16A10.1,

Catalog No. MABC1023 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC1023 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC1023 Supplier MilliporeSigma™ Supplier No. MABC1023
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects EGR1 clone: 16A10.1 in Human samples, and it is validated for Immunocytochemistry, Western Blotting

Early growth response protein 1 (UniProt P18146; also known as AT225, EGR-1, Nerve growth factor-induced protein A, NGFI-A, Transcription factor ETR103, Transcription factor Zif268, Zinc finger protein 225, Zinc finger protein Krox-24) is encoded by the EGR1 (also known as KROX24, ZNF225) gene (Gene ID 1958) in human. EGR-1 is the founding member of the early growth response (EGR) family of Cys2-His2-type zinc finger transcription factors that share a high homology in their DNA-binding region. EGR-1 is ubiquitously expressed in many cell types and binds to a consensus DNA motif (GCGGTGGGCG) in target genes, including c-Myc, cyclins D2 & G2, p19, TNF, IL-2, CD44, and ICAM-1. EGR1 is known as an immediate-early response gene due to the rapid kinetics of its induction by various signals, including growth factors, cytokines, and stress. Human EGR-1 is a 543-amino acid protein with three C2H2-type Zinc fingers (a.a. 338-362, 368-390, 396-418).
TRUSTED_SUSTAINABILITY

Specifications

Antigen EGR1
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 16A10.1
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P18146
Gene Symbols EGR1; KROX24; ZNF225
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to an internal fragment of human EGR1.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_001955
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.