Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EID1, Mouse, Clone: 2, Abnova™

Catalog No. 89113020 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-020 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-020 Supplier Abnova Corporation Supplier No. MAB6754

Mouse monoclonal antibody raised against synthetic peptide of EID1.

Specifications

Antigen EID1
Applications ELISA, Western Blot
Classification Monoclonal
Clone #2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of EID1.
Dilution Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol)
Gene EID1
Gene Alias C15orf3/CRI1/EID-1/IRO45620/MGC138883/MGC138884/PNAS-22/PTD014/RBP21
Gene Symbols EID1
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids 1-19 of human EID1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23741
Test Specificity This antibody reacts with human EID1.
Target Species Human
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.