Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-ELK1, Clone: 7E10D5, 7E10E8, Abnova™

Catalog No. 89136017 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-017 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-017 Supplier Abnova Corporation Supplier No. MAB5580
Add to Cart
Add to Cart

Specifications

Classification Monoclonal
Clone 7E10D5, 7E10E8
Description Mouse monoclonal antibody raised against partial recombinant ELK1.
Dilution ELISA (1:10000) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide)
Immunogen Recombinant protein corresponding to ELK1.
Quantity 100 μg
Primary or Secondary Primary
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.