Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-EMMPRIN/CD147, Clone: CB43, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2445360.2MG
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunofluorescence
EMMPRIN/CD147 Monoclonal antibody specifically detects EMMPRIN/CD147 in Human samples. It is validated for Flow Cytometry, Immunofluorescence.Specifications
EMMPRIN/CD147 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence 0.5 - 1.0 ug/ml | |
5F7, basigin, basigin (Ok blood group), CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
CD147 is a transmembrane glycoprotein of the immunoglobulin superfamily. It is expressed more intensely on thymocytes than on mature peripheral blood T cells. CD147 is important in spermatogenesis, embryo implantation, neural network formation, and tumor progression. CD147 is involved in the regulation of matrix remodeling at the epidermal-dermal interface. It stimulates the production of interstitial collagenase, gelatinase A, stromelysin-1 and various metalloproteinases (MMPs) by fibroblasts. These enzymes, which are typically increased during tissue degradation and wound healing, are important factors in cancer invasion and metastasis. | |
Store at 4C. | |
IgM |
Flow Cytometry, Immunofluorescence | |
CB43 | |
Unconjugated | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
BSG | |
Native human leucocytes | |
0.2 mg | |
Cancer, Signal Transduction, Vision | |
682 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction