Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENO2, Mouse, Clone: NSE-P2, Abnova™

Catalog No. 89110733 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-733 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-733 Supplier Abnova Corporation Supplier No. MAB1963
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of ENO2.

This gene encodes one of the three enolase isoenzymes found in mammals. This isoenzyme, a homodimer, is found in mature neurons and cells of neuronal origin. A switch from alpha enolase to gamma enolase occurs in neural tissue during development in rats and primates. [provided by RefSeq

Specifications

Antigen ENO2
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone NSE-P2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of ENO2.
Dilution Western blot (1 to 2 ug/mL) Immunohistochemistry on formalin to fixed/paraffin to embedded tissues (1 to 5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.08% sodium azide)
Gene ENO2
Gene Alias NSE
Gene Symbols ENO2
Host Species Mouse
Immunogen A synthetic peptide corresponding to human ENO2.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2026
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.