Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENPP3, Mouse, PE, Clone: NP4D6, Abnova™

Catalog No. 89112905 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-905 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-905 Supplier Abnova Corporation Supplier No. MAB6498
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native ENPP3.

The protein encoded by this gene belongs to a series of ectoenzymes that are involved in hydrolysis of extracellular nucleotides. These ectoenzymes possess ATPase and ATP pyrophosphatase activities and are type II transmembrane proteins. Expression of the related rat mRNA has been found in a subset of immature glial cells and in the alimentary tract. The corresponding rat protein has been detected in the pancreas, small intestine, colon, and liver. The human mRNA is expressed in glioma cells, prostate, and uterus. Expression of the human protein has been detected in uterus, basophils, and mast cells. [provided by RefSeq

Specifications

Antigen ENPP3
Applications Flow Cytometry
Classification Monoclonal
Clone NP4D6
Conjugate PE
Description Mouse monoclonal antibody raised against native ENPP3.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.2% BSA, 0.09% sodium azide)
Gene ENPP3
Gene Alias B10/CD203c/NPP3/PD-IBETA/PDNP3
Gene Symbols ENPP3
Host Species Mouse
Immunogen Native purified from HEK293 cells transfected with human ENPP3.
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5169
Test Specificity This antibody reacts with CD203C, a transmembrane ectoenzyme expressed on basophils and mast cells, and overexpressed upon their activation.
Target Species Human
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.