Learn More
EP300, Mouse, Clone: RW105, Abnova™
Mouse monoclonal antibody raised against partial recombinant EP300.
Supplier: Abnova Corporation MAB2385
Description
This gene encodes the adenovirus E1A-associated cellular p300 transcriptional co-activator protein. It functions as histone acetyltransferase that regulates transcription via chromatin remodeling and is important in the processes of cell proliferation and differentiation. It mediates cAMP-gene regulation by binding specifically to phosphorylated CREB protein. This gene has also been identified as a co-activator of HIF1A (hypoxia-inducible factor 1 alpha), and thus plays a role in the stimulation of hypoxia-induced genes such as VEGF. Defects in this gene are a cause of Rubinstein-Taybi syndrome and may also play a role in epithelial cancer. [provided by RefSeq
Specifications
| EP300 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| EP300 | |
| EP300 | |
| Recombinant protein corresponding to amino acids 1572-2371 of human EP300. | |
| RUO | |
| 2033 | |
| Human, Mouse, Primate, Rat | |
| Liquid |
| Immunofluorescence, Immunoprecipitation, Western Blot | |
| RW105 | |
| Mouse monoclonal antibody raised against partial recombinant EP300. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| KAT3B/p300 | |
| Mouse | |
| 200 μL | |
| Primary | |
| This antibody is specific to p300 protein. This antibody recognizes residues 1921-2023. | |
| Store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.