Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EP300, Mouse, Clone: RW128, Abnova™

Catalog No. 89110979 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-979 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-979 Supplier Abnova Corporation Supplier No. MAB2384
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant EP300.

This gene encodes the adenovirus E1A-associated cellular p300 transcriptional co-activator protein. It functions as histone acetyltransferase that regulates transcription via chromatin remodeling and is important in the processes of cell proliferation and differentiation. It mediates cAMP-gene regulation by binding specifically to phosphorylated CREB protein. This gene has also been identified as a co-activator of HIF1A (hypoxia-inducible factor 1 alpha), and thus plays a role in the stimulation of hypoxia-induced genes such as VEGF. Defects in this gene are a cause of Rubinstein-Taybi syndrome and may also play a role in epithelial cancer. [provided by RefSeq

Specifications

Antigen EP300
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone RW128
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant EP300.
Dilution Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene EP300
Gene Alias KAT3B/p300
Gene Symbols EP300
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1572-2371 of human EP300.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2033
Test Specificity This antibody is specific to p300 protein. This antibody recognizes residues 1868-1921.
Target Species Human, Mouse, Primate, Rat
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.