Learn More
EPB41L1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant EPB41L1.
Supplier: Abnova Corporation H00002036A01
Description
Erythrocyte membrane protein band 4.1 (EPB41) is a multifunctional protein that mediates interactions between the erythrocyte cytoskeleton and the overlying plasma membrane. The protein encoded by this gene is a neuronally-enriched protein that is structurally similar to EPB41. The encoded protein binds and stabilizes D2 and D3 dopamine receptors at the neuronal plasma membrane. Multiple transcript variants encoding different isoforms have been found for this gene, but the full-length nature of only two of them has been determined. [provided by RefSeq
Sequence: MEEKDYSEADGLSERTTPSKAQKSPQKIAKKYKSAICRVTLLDASEYECEVEKHGRGQVLFDLVCEHLNLLEKDYFGLTFCDADSQKNSpecifications
EPB41L1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant EPB41L1. | |
EPB41L1 | |
4.1N/DKFZp686H17242/KIAA0338/MGC11072 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_177996 | |
EPB41L1 | |
EPB41L1 (NP_818932, 1 a.a. to 88 a.a) partial recombinant protein with GST tag. | |
RUO | |
2036 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only