Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPCAM, Mouse, Clone: GZ-20, Abnova™

Catalog No. 89109396 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-396 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-396 Supplier Abnova Corporation Supplier No. MAB11225

Mouse monoclonal antibody raised against Epcam.

Specifications

Antigen EPCAM
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone GZ-20
Conjugate Unconjugated
Description tumor-associated calcium signal transducer 1
Formulation Bioreactor concentrate (0.05% sodium azide)
Gene Tacstd1
Gene Alias Egp314
Gene Symbols Tacstd1
Host Species Mouse
Immunogen Rat small intestine cells
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171577
Target Species Rat
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.