Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-EPHA2, Clone: 1B3C7, Abnova™

Catalog No. 89108831 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-831 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-831 Supplier Abnova Corporation Supplier No. MAB10635
Add to Cart
Add to Cart

Specifications

Classification Monoclonal
Clone 1B3C7
Description Mouse monoclonal antibody raised against partial recombinant EPHA2.
Dilution ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) The optimal working dilution should be determined by the end user.
Formulation In ascites (0.03% sodium azide)
Gene Alias ECK
Gene Symbols EPHA2
Immunogen Recombinant protein corresponding to human EPHA2.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 1969
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.