Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPHA2, Mouse, Clone: 3D7, Abnova™

Catalog No. 89110613 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-613 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-613 Supplier Abnova Corporation Supplier No. MAB1769
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant EPHA2.

This gene belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. This gene encodes a protein that binds ephrin-A ligands. [provided by RefSeq

Specifications

Antigen EPHA2
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 3D7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant EPHA2.
Dilution Western Blot (1:500-1:2000) Immunohistochemistry (1:100-1:200) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene EPHA2
Gene Alias ECK
Gene Symbols EPHA2
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 559-976 of human EPHA2.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1969
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.