Learn More
EPOR, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant EPOR.
Supplier: Abnova Corporation H00002057A01
Description
The erythropoietin receptor is a member of the cytokine receptor family. Upon erythropoietin binding, the erythropoietin receptor activates Jak2 tyrosine kinase which activates different intracellular pathways including: Ras/MAP kinase, phosphatidylinositol 3-kinase and STAT transcription factors. The stimulated erythropoietin receptor appears to have a role in erythroid cell survival. Defects in the erythropoietin receptor may produce erythroleukemia and familial erythrocytosis. [provided by RefSeq
Sequence: PDPKFESKAALLAARGPEELLCFTERLEDLVCFWEEAASAGVGPGNYSFSYQLEDEPWKLCRLHQAPTARGAVRFWCSLPTADTSSFVPLELRVTAASGASpecifications
EPOR | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant EPOR. | |
EPOR | |
MGC138358 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_000121 | |
EPOR | |
EPOR (NP_000112, 31 a.a. to 130 a.a) partial recombinant protein with GST tag. | |
RUO | |
2057 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only