Learn More
EPS8L1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant EPS8L1.
Supplier: Abnova Corporation H00054869A01
Description
This gene encodes a protein that is related to epidermal growth factor receptor pathway substrate 8 (EPS8), a substrate for the epidermal growth factor receptor. The function of this protein is unknown. At least two alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: RSPAAETPPLQRRPSVRAVISTVERGAGRGRPQAKPIPEAEEAQRPEPVGTSSNADSASPDLGPRGPDLAVLQAEREVDILNHVFDDVESFVSRLQKSASpecifications
EPS8L1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant EPS8L1. | |
EPS8L1 | |
DRC3/EPS8R1/FLJ20258/MGC23164/MGC4642/PP10566 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_133180 | |
EPS8L1 | |
EPS8L1 (NP_573441, 181 a.a. to 279 a.a) partial recombinant protein with GST tag. | |
RUO | |
54869 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only