Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epstein-Barr virus nuclear, Mouse, Clone: E1-2.5, Abnova™

Catalog No. 89136336 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-336 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-336 Supplier Abnova Corporation Supplier No. MAB7342

Mouse monoclonal antibody raised against synthetic pepetide of Epstein-Barr virus nuclear antigen.

Specifications

Antigen Epstein-Barr virus nuclear
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone E1-2.5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic pepetide of Epstein-Barr virus nuclear antigen.
Dilution ELISA (1:2000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen A synthetic peptide corresponding to repetitive Gly-Ala region of Epstein-Barr virus.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes repetitive gly-ala region of the nuclear antigen 1 of Epstein and Barr virus (EBNA-1). Not tested in other applications.
Target Species Epstein-Barr virus (EBV)
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.