Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERCC4, Mouse, Clone: 219, Abnova™

Catalog No. 89111385 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-385 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-385 Supplier Abnova Corporation Supplier No. MAB3381

Mouse monoclonal antibody raised against recombinant ERCC4.

The protein encoded by this gene forms a complex with ERCC1 and is involved in the 5' incision made during nucleotide excision repair. This complex is a structure specific DNA repair endonuclease that interacts with EME1. Defects in this gene are a cause of xeroderma pigmentosum complementation group F (XP-F), or xeroderma pigmentosum VI (XP6)

Specifications

Antigen ERCC4
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 219
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant ERCC4.
Dilution Western Blot (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) ( 2-4 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.4 (0.09% sodium azide)
Gene ERCC4
Gene Alias RAD1/XPF
Gene Symbols ERCC4
Host Species Mouse
Immunogen Recombinant protein corresponding to human ERCC4.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2072
Test Specificity This antibody recognizes a protein of approximately 110 KDa which is identified as XPF.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG2 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.