Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ESR1, Mouse, Clone: 5D4B1, 5D4E3, Abnova™

Catalog No. 89113428 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-428 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 89-113-428 Supplier Abnova Corporation Supplier No. MAB8304
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse monoclonal antibody raised against partial recombinant ESR1.

This gene encodes an estrogen receptor, a ligand-activated transcription factor composed of several domains important for hormone binding, DNA binding, and activation of transcription. The protein localizes to the nucleus where it may form a homodimer or a heterodimer with estrogen receptor 2. Estrogen and its receptors are essential for sexual development and reproductive function, but also play a role in other tissues such as bone. Estrogen receptors are also involved in pathological processes including breast cancer, endometrial cancer, and osteoporosis. Alternative splicing results in several transcript variants, which differ in their 5' UTRs and use different promoters. [provided by RefSeq]

Specifications

Antigen ESR1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5D4B1, 5D4E3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant ESR1.
Dilution Western Blot (1:500-1:2000) ELISA (1:10000) The optimal working dilution should be determined by the end user.
Formulation In ascites
Gene ESR1
Gene Alias DKFZp686N23123/ER/ESR/ESRA/Era/NR3A1
Gene Symbols ESR1
Host Species Mouse
Immunogen Recombinant protein corresponding to truncated human ESR1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2099
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.