Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Esrp1/Esrp2, Mouse, Clone: 23A7.C9, Abnova™

Catalog No. 89108465 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-465 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-465 Supplier Abnova Corporation Supplier No. MAB10272
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant Esrp1/Esrp2.

Specifications

Antigen Esrp1/Esrp2
Applications Western Blot
Classification Monoclonal
Clone 23A7.C9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant Esrp1/Esrp2.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene Alias Rbm35a/BC031468/A630065D16/RP23-322G9.1/2210008M09Rik
Gene Symbols Esrp1
Host Species Mouse
Immunogen Recombinant GST fusion protein corresponding to full length mouse Esrp1/Esrp2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 207920/77411
Test Specificity This antibody reacts with both mouse Esrp-1 and Esrp-2 proteins. A BLAST analysis of the immunizing protein sequence shows 100% homology with Esrp-1 from mouse and a 91% sequence homology with Esrp-1 from human, pig, rat, opossum, horse, cattle, panda, dog, and chimpanzee . The binding epitope of this monoclonal antibody has not been mapped
Target Species Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.