Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Estrogen receptor beta 1, Mouse, Clone: PPG5/10, Abnova™

Catalog No. 89135941 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-941 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-941 Supplier Abnova Corporation Supplier No. MAB5345

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse monoclonal antibody raised against synthetic peptide of Estrogen receptor beta 1.

Specifications

Antigen Estrogen receptor beta 1
Applications Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone PPG5/10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of Estrogen receptor beta 1.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:5-1:50) The optimal working dilution should be determined by the end user.
Formulation In tissue culture supernatant (0.09% sodium azide)
Host Species Mouse
Immunogen A synthetic peptide corresponding to C-terminus of human Estrogen receptor beta 1 isoform.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes the oestrogen receptor beta 1 isoform. This antibody has been used for the immunohistochemical study of the expression of this isoform, showing positive nuclear staining in cells within the ovary, uterus, oviduct, breast, testis, prostate, bladder and male reproductive ducts. There are also reports of staining by this antibody of cells in the adenohypophysis, demonstrating cytoplasmic expression. These results await further characterisation. Clone PPG5/10 reacts with new world primate, primates, human.
Target Species Human, Non-human Primate, Primate
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.