Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-Estrogen Receptor, Clone: ER503, Azide Free, Novus Biologicals™

Mouse Monoclonal Antibody
$631.00
Specifications
Antigen | ER alpha/NR3A1 |
---|---|
Clone | ER503 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 0.5-1ug/ml, ELISA 1-5ug/ml for coating, Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1ug/ml |
Applications | Western Blot, ELISA, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP23332301
![]() |
Novus Biologicals
NBP2333230.1MG |
0.1 mg |
Each for $631.00
|
|
|||||
NBP23332302
![]() |
Novus Biologicals
NBP2333230.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
ER alpha/NR3A1 Monoclonal specifically detects ER alpha/NR3A1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
ER alpha/NR3A1 | |
1.0 mg/mL | |
Western Blot, ELISA, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
Mouse | |
Angiogenesis, Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Dendritic Cell Markers, GPCR, Neuroscience, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
10mM PBS - BSA free with No Preservative | |
ER, ER alpha, Era, ER-alpha, ESRESRA, Estradiol receptor, estrogen receptor, estrogen receptor 1, estrogen receptor alpha, estrogen receptor alpha delta 3*4,56,7*/819-2 isoform, estrogen receptor alpha delta 4 +49 isoform, estrogen receptor alpha delta 4*5,6,7*/654 isoform, NR3A1DKFZp686N23123, Nuclear receptor subfamily 3 group A member 1 | |
ESR1 | |
IgG1 | |
Protein G purified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
ER503 | |
Western Blot 0.5-1ug/ml, ELISA 1-5ug/ml for coating, Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
P03372 | |
2099 | |
Recombinant human Estrogen Receptor alpha protein (aa2-185) | |
Primary | |
Protein A purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title