Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ETS2 (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89104457 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-457 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-457 Supplier Abnova Corporation Supplier No. H00002114A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant ETS2.

ETS transcriptions factors, such as ETS2, regulate numerous genes and are involved in stem cell development, cell senescence and death, and tumorigenesis. The conserved ETS domain within these proteins is a winged helix-turn-helix DNA-binding domain that recognizes the core consensus DNA sequence GGAA/T of target genes (Dwyer et al., 2007 [PubMed 17986575]).[supplied by OMIM

Sequence: YEENSHLTSVPHWINSNTLGFGTEQAPYGMQTQNYPKGGLLDSMCPASTPSVLSSEQEFQMFPKSRLSSVSVTYCSVSQDFPGSNLNLLTNNSGTPKDHD

Specifications

Antigen ETS2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant ETS2.
Formulation 50% glycerol
Gene ETS2
Gene Accession No. BC042954
Gene Alias ETS2IT1
Gene Symbols ETS2
Host Species Mouse
Immunogen ETS2 (AAH42954, 178 a.a. to 277 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2114
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.