Learn More
EVC, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant EVC.
Supplier: Abnova Corporation H00002121A01
Description
This gene encodes a protein containing a leucine zipper and a transmembrane domain. This gene has been implicated in both Ellis-van Creveld syndrome (EvC) and Weyers acrodental dysostosis. [provided by RefSeq
Sequence: VLERQRLMQCDLEEEENVRATEAVVALCQELYFSTVDTFQKFVDALFLQTLPGMTGLPPEECDYLRQEVQENAAWQLGKSNRFRRQQWKLFQELLEQDQQVWMEECALSSSpecifications
EVC | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant EVC. | |
EVC | |
DWF-1/EVC1/EVCL/MGC105107 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_014556 | |
EVC | |
EVC (NP_055371, 493 a.a. to 602 a.a) partial recombinant protein with GST tag. | |
RUO | |
2121 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only