Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EVI2B, Mouse, PE, Clone: MEM-216, Abnova™

Catalog No. 89113795 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-795 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-795 Supplier Abnova Corporation Supplier No. MAB9803
Add to Cart
Add to Cart

Mouse monoclonal antibody raised agaisnt full-length native EVI2B.

Specifications

Antigen EVI2B
Applications Flow Cytometry
Classification Monoclonal
Clone MEM-216
Conjugate PE
Description Mouse monoclonal antibody raised agaisnt full length native EVI2B.
Dilution Flow Cytometry (20 uL reagent/100 uL of whole blood or 106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.02% BSA, 0.09% sodium azide)
Gene EVI2B
Gene Alias D17S376/EVDB
Gene Symbols EVI2B
Host Species Mouse
Immunogen Native purified EVI2B from human Raji cells.
Purification Method Size-exclusion chromatography purification
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2124
Test Specificity The mouse monoclonal antibody MEM-216 recognizes EVI2B (CD361), almost uncharacterized type I transmembrane protein with broad leukocyte expression, mostly in myeloid and B cells.
Target Species Human
Content And Storage Store in the dark at 4°C. Avoid prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.