Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Exo1, Mouse, Clone: 266, Abnova™

Catalog No. 89111401 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-401 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-401 Supplier Abnova Corporation Supplier No. MAB3409

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse monoclonal antibody raised against full-length recombinant Exo1.

Specifications

Antigen Exo1
Applications Western Blot
Classification Monoclonal
Clone 266
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant Exo1.
Dilution Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.4 (0.09% sodium azide)
Gene Exo1
Gene Alias 5730442G03Rik/Msa
Gene Symbols Exo1
Host Species Mouse
Immunogen Recombinant protein corresponding to full length mouse Exo1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26909
Test Specificity Recognizes a protein of ∼92 KDa identified as the 5' to 3' double-strand DNA exonuclease (Exo1).
Target Species Human, Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.