Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EZR, Mouse, Clone: 45j8, Abnova™

Catalog No. 89113596 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-596 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-596 Supplier Abnova Corporation Supplier No. MAB8812
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant EZR.

The cytoplasmic peripheral membrane protein encoded by this gene functions as a protein-tyrosine kinase substrate in microvilli. As a member of the ERM protein family, this protein serves as an intermediate between the plasma membrane and the actin cytoskeleton. This protein plays a key role in cell surface structure adhesion, migration and organization, and it has been implicated in various human cancers. A pseudogene located on chromosome 3 has been identified for this gene. Alternatively spliced variants have also been described for this gene. [provided by RefSeq

Specifications

Antigen EZR
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 45j8
Concentration 1 ug/uL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant EZR.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.02% sodium azide)
Gene EZR
Gene Alias CVIL/CVL/DKFZp762H157/FLJ26216/MGC1584/VIL2
Gene Symbols EZR
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 362-585 at C-terminus of human EZR.
Purification Method In ascites
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7430
Target Species Bovine, Hamster, Human, Kangaroo, Monkey, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.