Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Mouse anti-Ezrin (Ascites Free), Clone: 4A5,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MAB3822C
Description
Specifically detects Ezrin (Ascites Free) clone: 4A5 in Human, Mouse, Rabbit, Rat samples, and it is validated for Immunohistochemistry, Immunoprecipitation, Western Blotting
Ezrin (UniProt Q8HZQ5; also known as Cytovillin, Villin-2, p81) is encoded by the EZR (also known as VIL2) gene (Gene ID 100008846) in Oryctolagus cuniculus (Rabbit) species. Ezrin, radixin, and moesin are the three ERM protein family members that crosslink actin filaments with plasma membranes and play an important role in the cell cortex architecture. In its inactive state, ezrin is located in the cytosol in a conformation involving intramolecular interaction between its N- and C-terminal. Ezrin is activated by phosphorylation, which causes a conformation change that disrupts the head-to-tail intramolecular interaction. Upon activation, ezrin binds to the cytoplasmic tails of multiple adhesion and signaling membrane molecules, such as CD44, growth factor receptors, and vascular adhesion molecule (VCAM1). The initiator methionine of ezrin is removed posttranslationally to yield the mature protein (a.a. 2-586) with an N-termina FERM domain (a.a. 2-295) that mediates interaction with microtubules.
Specifications
Ezrin (Ascites Free) | |
Monoclonal | |
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
EZR; VIL2 | |
Purified Ezrin from rabbit gastric parietal cells. | |
100 μg | |
Cell Structure | |
NP_001075591 | |
Stable for one year at 2°C to 8°C from date of receipt. | |
IgG1 κ |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
4A5 | |
Q8HZQ5 | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Human, Mouse, Rabbit, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction