Learn More
F11, Mouse, Monoclonal Antibody, Abnova™
Mouse monoclonal antibody raised against native F11.
Supplier: Abnova Corporation MAB0272
Description
This gene encodes coagulation factor XI of the blood coagulation cascade. This protein is present in plasma as a zymogen, which is a unique plasma coagulation enzyme because it exists as a homodimer consisting of two identical polypeptide chains linked by disulfide bonds. During activation of the plasma factor XI, an internal peptide bond is cleaved by factor XIIa (or XII) in each of the two chains, resulting in activated factor XIa, a serine protease composed of two heavy and two light chains held together by disulfide bonds. This activated plasma factor XI triggers the middle phase of the intrisic pathway of blood coagulation by activating factor IX. Defects in this factor lead to Rosenthal syndrome, a blood coagulation abnormality. [provided by RefSeq
Specifications
F11 | |
Monoclonal | |
Mouse monoclonal antibody raised against native F11. | |
In PBS (50% glycerol) | |
FXI/MGC141891 | |
Mouse | |
100 μg | |
Primary | |
This antibody recognizes human factor XI, 125I-factor XI, human factor XIa, BEGRck-factor XIa. It will inhibits clotting. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
ELISA, Radioimmune Assays (RIA), Western Blot | |
Unconjugated | |
Western Blot (5 ug/mL) The optimal working dilution should be determined by the end user. | |
F11 | |
F11 | |
Native purified Human F11. | |
RUO | |
2160 | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.