Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F5 Mouse anti-Bovine, Monoclonal Antibody, Abnova™

Catalog No. 89107893 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-893 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-893 Supplier Abnova Corporation Supplier No. MAB0259
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native F5.

Specifications

Antigen F5
Applications ELISA, Inhibition Assays, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native F5.
Dilution Western Blot (5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol)
Gene F5
Gene Symbols F5
Host Species Mouse
Immunogen Native purified Bovine F5.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 280687
Test Specificity This antibody recognizes bovine factor Va heavy chain, weakly with intact bovine factor V.
Target Species Bovine
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.