Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F7, Mouse, Monoclonal Antibody, Abnova™

Catalog No. 89107897 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-897 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-897 Supplier Abnova Corporation Supplier No. MAB0263
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native F7.

This gene encodes coagulation factor VII which is a vitamin K-dependent factor essential for hemostasis. This factor circulates in the blood in a zymogen form, and is converted to an active form by either factor IXa, factor Xa, factor XIIa, or thrombin by minor proteolysis. Upon activation of the factor VII, a heavy chain containing a catalytic domain and a light chain containing 2 EGF-like domains are generated, and two chains are held together by a disulfide bond. In the presence of factor III and calcium ions, the activated factor then further activates the coagulation cascade by converting factor IX to factor IXa and/or factor X to factor Xa. Alternative splicing of this gene results in 2 transcripts. Defects in this gene can cause coagulopathy. [provided by RefSeq

Specifications

Antigen F7
Applications ELISA, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native F7.
Dilution Western Blot (5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol)
Gene F7
Gene Symbols F7
Host Species Mouse
Immunogen Native purified Human F7.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2155
Test Specificity This antibody recognizes human factor VII, factor VIIa, BFPRck factor VIIa.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.