Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F8, Mouse, Clone: 101, Abnova™

Catalog No. 89111720 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-720 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-720 Supplier Abnova Corporation Supplier No. MAB4180
Add to Cart
Add to Cart
Request a Quote

Mouse monoclonal antibody raised against F8.

This gene encodes coagulation factor VIII, which participates in the intrinsic pathway of blood coagulation; factor VIII is a cofactor for factor IXa which, in the presence of Ca+2 and phospholipids, converts factor X to the activated form Xa. This gene produces two alternatively spliced transcripts. Transcript variant 1 encodes a large glycoprotein, isoform a, which circulates in plasma and associates with von Willebrand factor in a noncovalent complex. This protein undergoes multiple cleavage events. Transcript variant 2 encodes a putative small protein, isoform b, which consists primarily of the phospholipid binding domain of factor VIIIc. This binding domain is essential for coagulant activity. Defects in this gene results in hemophilia A, a common recessive X-linked coagulation disorder. [provided by RefSeq]

Specifications

Antigen F8
Applications ELISA, Western Blot
Classification Monoclonal
Clone 101
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against F8.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4
Gene F8
Gene Alias AHF/DXS1253E/F8B/F8C/FVIII/HEMA
Gene Symbols F8
Host Species Mouse
Immunogen Purified human F8.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2157
Test Specificity Recognizes full-length human Factor VIII. Does not cross-react with von Willebrand factor.
Target Species Human
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.