Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FABP4, Mouse, Clone: 3F4, Abnova™

Catalog No. 89110800 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-800 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-800 Supplier Abnova Corporation Supplier No. MAB2059
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant FABP4.

FABP4 encodes the fatty acid binding protein found in adipocytes. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. It is thought that FABPs roles include fatty acid uptake, transport, and metabolism. [provided by RefSeq

Specifications

Antigen FABP4
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3F4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant FABP4.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene FABP4
Gene Alias A-FABP/aP2
Gene Symbols FABP4
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-132 of human FABP4.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2167
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.