Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAH, Mouse, Clone: 665a3, Abnova™

Catalog No. 89113578 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-578 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-578 Supplier Abnova Corporation Supplier No. MAB8773
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant FAH.

This gene encodes the last enzyme in the tyrosine catabolism pathway. FAH deficiency is associated with Type 1 hereditary tyrosinemia (HT). [provided by RefSeq

Specifications

Antigen FAH
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 665a3
Concentration 1 ug/uL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant FAH.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In citrate-Tris-HCl, pH 7.0 (0.02% Proclin 300)
Gene FAH
Gene Symbols FAH
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human FAH.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2184
Test Specificity This antibody recognizes FAH expression in HEK293 whole cell lysate.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.