Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM119B, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106669 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-669 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-669 Supplier Abnova Corporation Supplier No. H00025895B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human FAM119B protein.

Sequence: MADPGPDPESESESVFPREVGLFADSYSEKSQFCFCGHVLTITQNFGSRLGVAARVWDAALSLCNYFESQNVDFRGKKVIELGAGTGIVGILAALQGGDVTITDLPLALEQIQGNVQANVPAGGQAQVRALSWGIDHHVFPANYDLVLGADIVYLEPTFPLLLGTLQHLCRPHGTIYLASKMRKEHGTESFFQHLLPQHFQLELAQRDEDENVNIYRARHREPRPA

Specifications

Antigen FAM119B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human FAM119B protein.
Formulation PBS with no preservative; pH 7.4
Gene FAM119B
Gene Accession No. NM_015433.2
Gene Alias DKFZp586D0919
Gene Symbols FAM119B
Host Species Mouse
Immunogen FAM119B (NP_056248.2,1 a.a. ∽ 226 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25895
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.