Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM12A, Mouse, Clone: 3D9, Abnova™

Catalog No. 89106419 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-419 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-419 Supplier Abnova Corporation Supplier No. H00010876M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant FAM12A.

Testicular sperm are morphologically differentiated but are not progressively motile nor able to fertilize an egg. Post-testicular maturation requires exposure of spermatozoa to the microenvironment of the epididymal lumen. Spermatozoa undergo extensive changes in the epididymis, including enzymatic modifications, loss of pre-existing components and addition of new glycoproteins from epididymal secretions. These modifying proteins and enzymes are synthesized by epithelial cells lining the epididymal duct and secreted apically into the lumen, where they come into contact with, and may be absorbed onto, the sperm membranes. The proteins encoded by the genes in this cluster are synthesized and secreted by epididymal epithelial cells. [provided by RefSeq

Sequence: MTSSLKIWGILLALLCILCRLCVYSNNIYWREFIKLHYLSPSREFKEYKCDVLMREKEALKGKSFHMFIYSLWFKIQRACINEKGSDRYRNAYVWAPGALKVLECHWEKYNNRYTESRSFSYIEFHCGVDGYVDNIEDLRIIEPISN

Specifications

Antigen FAM12A
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3D9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant FAM12A.
Formulation PBS with no preservative; pH 7.4
Gene FAM12A
Gene Accession No. NM_006683.4
Gene Alias EP3A/HE3-ALPHA/HE3A/HE3ALPHA/MGC119614/MGC119615
Gene Symbols FAM12A
Host Species Mouse
Immunogen FAM12A (NP_006674.2,1 a.a. ∽ 147 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A Purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10876
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.