Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM175A, Mouse, Clone: ABRA1-01, Abnova™

Catalog No. 89113477 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-477 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-477 Supplier Abnova Corporation Supplier No. MAB8508
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant FAM175A.

Specifications

Antigen FAM175A
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone ABRA1-01
Concentration 1 mg/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant FAM175A.
Dilution Flow cytometry (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene FAM175A
Gene Alias ABRA1/CCDC98/FLJ11520/FLJ12642/FLJ13614
Gene Symbols FAM175A
Host Species Mouse
Immunogen Recombinant His fusion protein corresponding to amino acids 1-313 of human FAM175A.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84142
Test Specificity This antibody is spevific to N-terminal part of FAM175A.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.