Learn More
Specifications
Specifications
| Classification | Monoclonal |
| Clone | FI-17 |
| Description | Mouse monoclonal antibody raised against recombinant FANCD2. |
| Dilution | ChIP (1:1000-1:2000) Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the end user. |
| Formulation | In buffer containing 0.09% sodium azide |
| Immunogen | Recombinant fusion protein corresponding to FANCD2. |
| Quantity | 100 μL |
| Primary or Secondary | Primary |
| Content And Storage | Store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.